Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DACH2
Entrez Gene ID
117154
Gene Name
dachshund family transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.X: 86148451 - 86832602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139514.1 NP_001132986.1 7 - 8 1520 64
NM_001139515.1 NP_001132987.1 8 - 9 945 64
NM_053281.3 NP_444511.1 8 - 9 1559 64
XM_011530848.2 XP_011529150.1 8 - 9 1558 64
XM_017029255.1 XP_016884744.1 7 - 8 1519 64
XM_017029256.1 XP_016884745.1 7 - 8 1519 64
GenBank mRNA AF428101.1 - 8 - 9 1389 64
AK055953.1 - 7 - 8 906 64
AK091842.1 - 7 - 8 1513 64
AK290596.1 - 8 - 9 1559 64
AK295648.1 - 3 - 4 355 64
AK298787.1 - 8 - 9 945 64
AL834456.1 - 1 - 2 151 64
BC114950.1 - 8 - 9 1468 64

View Full Product Details