Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL16
Entrez Gene ID
3603
Gene Name
interleukin 16
Gene Aliases
LCF, NIL16, PRIL16, prIL-16
Chromosome Location
Chr.15: 81181844 - 81312763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL833554.1 - 1 - 2 36 75
CR749286.1 - 1 - 2 34 75

View Full Product Details