Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSF1
Entrez Gene ID
51773
Gene Name
remodeling and spacing factor 1
Gene Aliases
HBXAP, RSF-1, XAP8, p325
Chromosome Location
Chr.11: 77660009 - 77872232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016578.3 NP_057662.3 9 - 10 3022 107
XM_005274051.2 XP_005274108.1 9 - 10 3048 107
XM_017017923.1 XP_016873412.1 9 - 10 2796 107
GenBank mRNA AF227948.2 - 4 - 5 3207 107
AF380176.1 - 9 - 10 2893 107
AK001268.1 - 4 - 5 1444 107
BC046124.1 - 5 - 6 2305 107

View Full Product Details