The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016578.3 | NP_057662.3 | 11 - 12 | 3185 | 98 |
XM_005274051.2 | XP_005274108.1 | 11 - 12 | 3211 | 98 | |
XM_017017923.1 | XP_016873412.1 | 11 - 12 | 2959 | 98 | |
GenBank mRNA | AF227948.2 | - | 6 - 7 | 3370 | 98 |
AF380176.1 | - | 11 - 12 | 3056 | 98 | |
AK001268.1 | - | 6 - 7 | 1607 | 98 |