The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190995.1 | NP_001177924.1 | 6 - 7 | 851 | 121 |
NM_006390.3 | NP_006381.2 | 10 - 11 | 1489 | 121 | |
XM_017018691.1 | XP_016874180.1 | 10 - 11 | 1386 | 121 | |
XM_017018692.1 | XP_016874181.1 | 9 - 10 | 1046 | 121 | |
XM_017018693.1 | XP_016874182.1 | 10 - 11 | 1482 | 121 | |
GenBank mRNA | AK302260.1 | - | 6 - 7 | 817 | 121 |
BX953637.1 | - | 4 - 5 | 351 | 121 | |
DB067822.1 | - | 3 - 4 | 287 | 121 | |
U77494.1 | - | 10 - 11 | 1311 | 121 |