The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190995.1 | NP_001177924.1 | 18 - 19 | 2407 | 88 |
NM_006390.3 | NP_006381.2 | 22 - 23 | 3045 | 88 | |
XM_017018691.1 | XP_016874180.1 | 22 - 23 | 2942 | 88 | |
XM_017018692.1 | XP_016874181.1 | 21 - 22 | 2602 | 88 | |
GenBank mRNA | AB209602.1 | - | 9 - 10 | 1125 | 88 |
AK130090.1 | - | 2 - 3 | 224 | 88 | |
AK302260.1 | - | 18 - 19 | 2373 | 88 | |
U77494.1 | - | 22 - 23 | 2867 | 88 |