Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IPO8
Entrez Gene ID
10526
Gene Name
importin 8
Gene Aliases
RANBP8
Chromosome Location
Chr.12: 30628981 - 30695995 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190995.1 NP_001177924.1 5 - 6 748 137
NM_006390.3 NP_006381.2 9 - 10 1386 137
XM_017018691.1 XP_016874180.1 9 - 10 1283 137
XM_017018692.1 XP_016874181.1 8 - 9 943 137
XM_017018693.1 XP_016874182.1 9 - 10 1379 137
GenBank mRNA AK302260.1 - 5 - 6 714 137
BX953637.1 - 3 - 4 248 137
DB067822.1 - 2 - 3 184 137
U77494.1 - 9 - 10 1208 137

View Full Product Details