The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318066.1 | NP_001304995.1 | 1 - 2 | 252 | 131 |
NM_014822.3 | NP_055637.2 | 1 - 2 | 252 | 131 | |
GenBank mRNA | AB018298.1 | - | 1 - 2 | 155 | 131 |
AK291765.1 | - | 1 - 2 | 152 | 131 | |
BC035761.1 | - | 1 - 2 | 174 | 131 | |
DA034453.1 | - | 1 - 2 | 253 | 131 | |
DB224337.1 | - | 1 - 2 | 227 | 131 |