The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005723.3 | NP_005714.2 | 1 - 2 | 523 | 95 |
XM_005262680.1 | XP_005262737.1 | 1 - 2 | 523 | 95 | |
GenBank mRNA | AF065389.1 | - | 1 - 2 | 435 | 95 |
AK315723.1 | - | 1 - 2 | 490 | 95 | |
BC009704.1 | - | 1 - 2 | 421 | 95 | |
CR541809.1 | - | 1 - 2 | 88 | 95 | |
CR541829.1 | - | 1 - 2 | 88 | 95 |