Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
Chromosome Location
Chr.1: 169511954 - 169586632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000130.4 NP_000121.2 23 - 24 6488 109
XM_017000660.1 XP_016856149.1 22 - 23 6497 109
GenBank mRNA AK226079.1 - 23 - 24 6406 109
M14335.1 - 23 - 24 6419 109
M16967.1 - 23 - 24 6433 109

View Full Product Details