The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000126.3 | NP_000117.1 | 4 - 5 | 434 | 68 |
NM_001127716.1 | NP_001121188.1 | 3 - 4 | 287 | 68 | |
GenBank mRNA | AK292979.1 | - | 4 - 5 | 434 | 68 |
AK300044.1 | - | 3 - 4 | 222 | 68 | |
BC015526.1 | - | 4 - 5 | 378 | 68 | |
BC029995.1 | - | 4 - 5 | 423 | 68 | |
BC095457.1 | - | 4 - 5 | 359 | 68 | |
BT009796.1 | - | 4 - 5 | 353 | 68 | |
BU176141.1 | - | 3 - 4 | 250 | 68 | |
DA838974.1 | - | 4 - 5 | 434 | 68 | |
J04058.1 | - | 4 - 5 | 353 | 68 | |
KU178046.1 | - | 4 - 5 | 353 | 68 | |
KU178047.1 | - | 5 - 6 | 486 | 68 | |
KU178048.1 | - | 4 - 5 | 353 | 68 |