Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
Chromosome Location
Chr.3: 46436005 - 46485234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199149.1 NP_001186078.1 16 - 17 2284 92
NM_001321121.1 NP_001308050.1 16 - 17 2389 92
NM_001321122.1 NP_001308051.1 19 - 20 2628 92
NM_002343.5 NP_002334.2 16 - 17 2395 92
GenBank mRNA AK290859.1 - 16 - 17 2140 92
AK292813.1 - 16 - 17 2140 92
AK298035.1 - 16 - 17 2284 92
AK303995.1 - 8 - 9 1003 92
AK316330.1 - 16 - 17 2204 92
AY165046.1 - 16 - 17 2114 92
AY493417.1 - 16 - 17 2212 92
BC015822.1 - 16 - 17 2144 92
BC015823.1 - 16 - 17 2146 92
BC022347.1 - 16 - 17 2140 92
EU794669.1 - 16 - 17 2137 92
KF992208.1 - 16 - 17 2114 92
M18642.1 - 6 - 7 797 92
M83202.1 - 16 - 17 2109 92
M83205.1 - 16 - 17 2088 92
M93150.1 - 16 - 17 2119 92
U07643.1 - 16 - 17 2199 92
X52941.1 - 16 - 17 2096 92
X53961.1 - 16 - 17 2396 92

View Full Product Details