Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMM50
Entrez Gene ID
25813
Gene Name
SAMM50 sorting and assembly machinery component
Gene Aliases
CGI-51, OMP85, SAM50, TOB55, TRG-3, YNL026W
Chromosome Location
Chr.22: 43955381 - 43996532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015380.4 NP_056195.3 5 - 6 622 81
GenBank mRNA AF151809.1 - 5 - 6 534 81
AK001087.1 - 5 - 6 561 81
AK222657.1 - 5 - 6 496 81
AK312773.1 - 5 - 6 457 81
AY189688.1 - 5 - 6 537 81
BC007830.1 - 5 - 6 529 81
BC011681.2 - 5 - 6 582 81
BC015200.1 - 5 - 6 472 81
CR456483.1 - 5 - 6 434 81

View Full Product Details