The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138814.3 | NP_620169.1 | 2 - 3 | 565 | 87 |
GenBank mRNA | AK056648.1 | - | 2 - 3 | 562 | 87 |
BC031820.2 | - | 2 - 3 | 529 | 87 | |
BC104839.1 | - | 2 - 3 | 479 | 87 | |
BC104843.1 | - | 2 - 3 | 479 | 87 |