The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001156.4 | NP_001147.1 | 7 - 8 | 707 | 117 |
NM_001320879.1 | NP_001307808.1 | 6 - 7 | 652 | 117 | |
NM_001320880.1 | NP_001307809.1 | 7 - 8 | 718 | 117 | |
NM_004034.3 | NP_004025.1 | 8 - 9 | 773 | 117 | |
XM_017016162.1 | XP_016871651.1 | 8 - 9 | 809 | 117 | |
XM_017016163.1 | XP_016871652.1 | 7 - 8 | 743 | 117 | |
GenBank mRNA | AB062429.1 | - | 7 - 8 | 665 | 117 |
AK222552.1 | - | 7 - 8 | 675 | 117 | |
AK295319.1 | - | 7 - 8 | 584 | 117 | |
AK300086.1 | - | 5 - 6 | 474 | 117 | |
AK301681.1 | - | 7 - 8 | 461 | 117 | |
AK307772.1 | - | 6 - 7 | 526 | 117 | |
AK309141.1 | - | 7 - 8 | 469 | 117 | |
AK312447.1 | - | 8 - 9 | 746 | 117 | |
AK313024.1 | - | 7 - 8 | 650 | 117 | |
BC002632.2 | - | 7 - 8 | 667 | 117 | |
BT007187.1 | - | 6 - 7 | 632 | 117 | |
BX361177.2 | - | 6 - 7 | 618 | 117 | |
CR407686.1 | - | 6 - 7 | 632 | 117 | |
J04543.1 | - | 7 - 8 | 692 | 117 |