Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- COA1
- Entrez Gene ID
-
55744
- Gene Name
- cytochrome c oxidase assembly factor 1 homolog
- Gene Aliases
-
C7orf44, MITRAC15
- Chromosome Location
-
Chr.7: 43608452 - 43729541 on Build GRCh38
View Full Product Details