The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281932.1 | NP_001268861.1 | 12 - 13 | 1344 | 97 |
NM_001281933.1 | NP_001268862.1 | 11 - 12 | 1291 | 97 | |
NM_017760.6 | NP_060230.5 | 11 - 12 | 1291 | 97 | |
XM_005249547.2 | XP_005249604.1 | 11 - 12 | 1289 | 97 | |
XM_011516356.2 | XP_011514658.1 | 11 - 12 | 1289 | 97 | |
XM_011516357.2 | XP_011514659.1 | 12 - 13 | 1317 | 97 | |
XM_011516358.2 | XP_011514660.1 | 11 - 12 | 1289 | 97 | |
XM_017012374.1 | XP_016867863.1 | 11 - 12 | 1289 | 97 | |
XM_017012375.1 | XP_016867864.1 | 11 - 12 | 1289 | 97 | |
XM_017012376.1 | XP_016867865.1 | 11 - 12 | 1289 | 97 | |
GenBank mRNA | AK000318.1 | - | 6 - 7 | 549 | 97 |
AK092008.1 | - | 12 - 13 | 1279 | 97 | |
AK315699.1 | - | 9 - 10 | 908 | 97 | |
BC020560.1 | - | 6 - 7 | 615 | 97 | |
BC043404.1 | - | 11 - 12 | 1232 | 97 | |
BX537845.1 | - | 11 - 12 | 1258 | 97 |