The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281932.1 | NP_001268861.1 | 8 - 9 | 965 | 106 |
NM_001281933.1 | NP_001268862.1 | 7 - 8 | 912 | 106 | |
NM_017760.6 | NP_060230.5 | 7 - 8 | 912 | 106 | |
NR_104054.1 | 9 - 10 | 1066 | 106 | ||
XM_005249547.2 | XP_005249604.1 | 7 - 8 | 910 | 106 | |
XM_011516356.2 | XP_011514658.1 | 7 - 8 | 910 | 106 | |
XM_011516357.2 | XP_011514659.1 | 8 - 9 | 938 | 106 | |
XM_011516358.2 | XP_011514660.1 | 7 - 8 | 910 | 106 | |
XM_017012374.1 | XP_016867863.1 | 7 - 8 | 910 | 106 | |
XM_017012375.1 | XP_016867864.1 | 7 - 8 | 910 | 106 | |
XM_017012376.1 | XP_016867865.1 | 7 - 8 | 910 | 106 | |
XM_017012378.1 | XP_016867867.1 | 5 - 6 | 529 | 106 | |
GenBank mRNA | AK000318.1 | - | 2 - 3 | 170 | 106 |
AK092008.1 | - | 8 - 9 | 900 | 106 | |
AK315699.1 | - | 5 - 6 | 529 | 106 | |
BC020560.1 | - | 2 - 3 | 236 | 106 | |
BC043404.1 | - | 7 - 8 | 853 | 106 | |
BX537845.1 | - | 7 - 8 | 879 | 106 |