Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRK4
Entrez Gene ID
2868
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
GPRK2L, GPRK4, GRK4a, IT11
Chromosome Location
Chr.4: 2963504 - 3040760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004056.1 NP_001004056.1 2 - 3 616 69
NM_001004057.1 NP_001004057.1 3 - 4 712 69
NM_005307.2 NP_005298.2 2 - 3 616 69
NM_182982.2 NP_892027.2 3 - 4 712 69
XM_011513447.2 XP_011511749.1 3 - 4 824 69
XM_011513448.2 XP_011511750.1 3 - 4 824 69
XM_011513449.2 XP_011511751.1 2 - 3 728 69
XM_011513450.2 XP_011511752.1 3 - 4 824 69
XM_011513451.2 XP_011511753.1 3 - 4 824 69
XM_011513452.2 XP_011511754.1 3 - 4 824 69
XM_011513453.2 XP_011511755.1 3 - 4 824 69
XM_011513454.2 XP_011511756.1 3 - 4 824 69
XM_011513455.2 XP_011511757.1 3 - 4 824 69
XM_011513456.2 XP_011511758.1 3 - 4 824 69
XM_011513457.2 XP_011511759.1 3 - 4 824 69
XM_017008053.1 XP_016863542.1 3 - 4 824 69
XM_017008054.1 XP_016863543.1 3 - 4 824 69
XM_017008055.1 XP_016863544.1 2 - 3 728 69
XM_017008056.1 XP_016863545.1 3 - 4 824 69
XM_017008061.1 XP_016863550.1 2 - 3 479 69
XM_017008063.1 XP_016863552.1 2 - 3 479 69
XM_017008064.1 XP_016863553.1 2 - 3 263 69
XM_017008065.1 XP_016863554.1 3 - 4 824 69
GenBank mRNA AK223581.1 - 2 - 3 624 69
AK292390.1 - 3 - 4 757 69
BC117320.1 - 3 - 4 600 69
U33054.1 - 3 - 4 511 69
U33055.1 - 2 - 3 415 69
U33056.1 - 3 - 4 511 69
X97879.1 - 2 - 3 259 69
X97880.1 - 2 - 3 259 69
X97881.1 - 3 - 4 355 69
X98118.1 - 3 - 4 355 69

View Full Product Details