The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256030.1 | NP_001242959.1 | 2 - 3 | 471 | 124 |
NM_001778.3 | NP_001769.2 | 2 - 3 | 471 | 124 | |
XM_011510171.1 | XP_011508473.1 | 3 - 4 | 539 | 124 | |
GenBank mRNA | AK225636.1 | - | 2 - 3 | 444 | 124 |
AK310624.1 | - | 2 - 3 | 445 | 124 | |
AK310744.1 | - | 2 - 3 | 451 | 124 | |
BC016182.1 | - | 2 - 3 | 415 | 124 | |
BG505217.1 | - | 2 - 3 | 471 | 126 | |
BT019813.1 | - | 2 - 3 | 383 | 124 | |
CR457012.1 | - | 2 - 3 | 383 | 124 | |
M37766.1 | - | 2 - 3 | 403 | 124 | |
M59904.1 | - | 2 - 3 | 400 | 124 | |
X06341.1 | - | 2 - 3 | 419 | 124 |