Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED17
Entrez Gene ID
9440
Gene Name
mediator complex subunit 17
Gene Aliases
CRSP6, CRSP77, DRIP80, TRAP80
Chromosome Location
Chr.11: 93784239 - 93813330 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004268.4 NP_004259.3 11 - 12 2015 71
GenBank mRNA AF104254.1 - 7 - 8 936 71
AF105421.1 - 11 - 12 1961 71
AF117657.2 - 11 - 12 1935 71
AK001674.1 - 11 - 12 1972 71
AK023209.1 - 11 - 12 1929 71
AK299702.1 - 12 - 13 1882 71
BC021101.1 - 11 - 12 1955 71

View Full Product Details