The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004268.4 | NP_004259.3 | 9 - 10 | 1738 | 140 |
XM_011543068.2 | XP_011541370.1 | 9 - 10 | 1697 | 140 | |
GenBank mRNA | AF104254.1 | - | 5 - 6 | 659 | 140 |
AF105421.1 | - | 9 - 10 | 1684 | 140 | |
AF117657.2 | - | 9 - 10 | 1658 | 140 | |
AK001674.1 | - | 9 - 10 | 1695 | 140 | |
AK023209.1 | - | 9 - 10 | 1652 | 140 | |
AK299702.1 | - | 10 - 11 | 1605 | 140 | |
BC021101.1 | - | 9 - 10 | 1678 | 140 |