Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPD52L1
Entrez Gene ID
7164
Gene Name
tumor protein D52-like 1
Gene Aliases
D53
Chromosome Location
Chr.6: 125153483 - 125264407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003395.2 NP_001003395.1 4 - 5 706 113
NM_003287.3 NP_003278.1 4 - 5 604 113
XM_005267122.2 XP_005267179.1 4 - 5 385 113
XM_006715557.3 XP_006715620.1 4 - 5 850 113
XM_017011237.1 XP_016866726.1 4 - 5 850 113
GenBank mRNA AK291887.1 - 4 - 5 399 113
BM048527.1 - 4 - 5 624 112
U44427.1 - 4 - 5 561 113
U44428.1 - 1 - 2 67 113

View Full Product Details