The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021733.1 | NP_068379.1 | 4 - 5 | 592 | 70 |
XM_011527201.1 | XP_011525503.1 | 4 - 5 | 662 | 70 | |
XM_011527202.1 | XP_011525504.1 | 4 - 5 | 662 | 70 | |
XM_011527203.2 | XP_011525505.1 | 4 - 5 | 662 | 70 | |
GenBank mRNA | BC058862.1 | - | 4 - 5 | 620 | 70 |