The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019087.2 | NP_061960.1 | 1 - 2 | 134 | 85 |
GenBank mRNA | AK000058.1 | - | 1 - 2 | 134 | 85 |
AK290886.1 | - | 1 - 2 | 135 | 85 | |
BC026093.1 | - | 1 - 2 | 138 | 85 |