Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGS17
Entrez Gene ID
26575
Gene Name
regulator of G-protein signaling 17
Gene Aliases
RGS-17, RGSZ2, hRGS17
Chromosome Location
Chr.6: 153010891 - 153131284 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012419.4 NP_036551.3 1 - 2 127 137
XM_017010733.1 XP_016866222.1 1 - 2 157 137
GenBank mRNA AF202257.3 - 1 - 2 138 137
BC013117.1 - 1 - 2 122 137

View Full Product Details