Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SESN3
Entrez Gene ID
143686
Gene Name
sestrin 3
Gene Aliases
SEST3
Chromosome Location
Chr.11: 95165513 - 95232541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144665.3 NP_653266.2 1 - 2 301 78
GenBank mRNA AK096300.1 - 1 - 2 301 78
BC017296.2 - 1 - 2 265 78

View Full Product Details