The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303130.1 | NP_001290059.1 | 7 - 8 | 800 | 148 |
NM_001303131.1 | NP_001290060.1 | 8 - 9 | 1287 | 148 | |
NM_174942.2 | NP_777602.1 | 8 - 9 | 921 | 148 | |
XM_005268804.3 | XP_005268861.1 | 9 - 10 | 990 | 148 | |
XM_011538219.2 | XP_011536521.1 | 8 - 9 | 846 | 148 | |
XM_011538220.2 | XP_011536522.1 | 7 - 8 | 2181 | 148 | |
XM_011538221.2 | XP_011536523.1 | 7 - 8 | 1031 | 148 | |
XM_017019206.1 | XP_016874695.1 | 9 - 10 | 1064 | 148 | |
GenBank mRNA | AK095594.1 | - | 8 - 9 | 1287 | 148 |
AK315189.1 | - | 7 - 8 | 778 | 148 | |
BC043366.1 | - | 8 - 9 | 871 | 148 |