Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MTHFD1L
- Entrez Gene ID
-
25902
- Gene Name
- methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like
- Gene Aliases
-
FTHFSDC1, MTC1THFS, dJ292B18.2
- Chromosome Location
-
Chr.6: 150865679 - 151101887 on Build GRCh38
View Full Product Details