The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005865.3 | NP_005856.1 | 11 - 12 | 1487 | 84 |
XM_017010161.1 | XP_016865650.1 | 10 - 11 | 1409 | 84 | |
XM_017010162.1 | XP_016865651.1 | 10 - 11 | 1307 | 84 | |
XM_017010163.1 | XP_016865652.1 | 10 - 11 | 1196 | 84 | |
XM_017010164.1 | XP_016865653.1 | 9 - 10 | 1016 | 84 | |
XM_017010165.1 | XP_016865654.1 | 5 - 6 | 738 | 84 | |
GenBank mRNA | AF052514.1 | - | 11 - 12 | 1484 | 84 |
AK093590.1 | - | 2 - 3 | 1376 | 84 | |
AK097918.1 | - | 5 - 6 | 738 | 84 | |
AK098033.1 | - | 2 - 3 | 663 | 84 | |
AK126160.1 | - | 2 - 3 | 2029 | 84 | |
AK226091.1 | - | 2 - 3 | 507 | 84 | |
AK303106.1 | - | 6 - 7 | 808 | 84 | |
AK314037.1 | - | 11 - 12 | 1487 | 84 |