The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005865.3 | NP_005856.1 | 8 - 9 | 1016 | 86 |
XM_017010161.1 | XP_016865650.1 | 7 - 8 | 938 | 86 | |
XM_017010162.1 | XP_016865651.1 | 8 - 9 | 1016 | 86 | |
GenBank mRNA | AF052514.1 | - | 8 - 9 | 1013 | 86 |
AK314037.1 | - | 8 - 9 | 1016 | 86 |