The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005865.3 | NP_005856.1 | 9 - 10 | 1164 | 69 |
XM_017010161.1 | XP_016865650.1 | 8 - 9 | 1086 | 69 | |
XM_017010163.1 | XP_016865652.1 | 8 - 9 | 873 | 69 | |
XM_017010165.1 | XP_016865654.1 | 3 - 4 | 415 | 69 | |
GenBank mRNA | AF052514.1 | - | 9 - 10 | 1161 | 69 |
AK097918.1 | - | 3 - 4 | 415 | 69 | |
AK303106.1 | - | 4 - 5 | 485 | 69 | |
AK314037.1 | - | 9 - 10 | 1164 | 69 |