The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017768.4 | NP_060238.3 | 13 - 14 | 1649 | 130 |
XM_005271013.1 | XP_005271070.1 | 12 - 13 | 1476 | 130 | |
XM_011541763.1 | XP_011540065.1 | 11 - 12 | 1391 | 130 | |
XM_017001695.1 | XP_016857184.1 | 12 - 13 | 1397 | 130 | |
XM_017001696.1 | XP_016857185.1 | 10 - 11 | 1171 | 130 | |
GenBank mRNA | AK000338.1 | - | 11 - 12 | 1159 | 130 |
AK022951.1 | - | 13 - 14 | 1563 | 130 | |
AK225337.1 | - | 11 - 12 | 1159 | 130 | |
AL162076.1 | - | 3 - 4 | 224 | 130 | |
BC003407.1 | - | 13 - 14 | 1449 | 130 | |
BC008586.1 | - | 13 - 14 | 1547 | 130 |