Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
Chromosome Location
Chr.2: 237324012 - 237434822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004369.3 NP_004360.2 28 - 29 7164 79
NM_057166.4 NP_476507.3 25 - 26 5343 79
NM_057167.3 NP_476508.2 27 - 28 6546 79
XM_005246065.1 XP_005246122.1 27 - 28 6521 79
XM_005246066.1 XP_005246123.1 25 - 26 5667 79
XM_006712253.1 XP_006712316.1 28 - 29 6620 79
XM_011510574.1 XP_011508876.1 28 - 29 7118 79
XM_017003303.1 XP_016858792.1 33 - 34 7171 79
XM_017003304.1 XP_016858793.1 30 - 31 5383 79
GenBank mRNA AB209636.1 - 19 - 20 2455 79
BC033174.1 - 5 - 6 287 79
BC144595.1 - 25 - 26 5284 79
BC150625.1 - 25 - 26 5284 79
BC171790.1 - 25 - 26 5284 79
BX641155.1 - 5 - 6 1133 79
M20778.1 - 14 - 15 768 79
X52022.1 - 28 - 29 7134 79

View Full Product Details