Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
Chromosome Location
Chr.2: 237324012 - 237434822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004369.3 NP_004360.2 5 - 6 2180 90
NM_057164.4 NP_476505.3 3 - 4 959 90
NM_057165.4 NP_476506.3 4 - 5 1562 90
NM_057167.3 NP_476508.2 4 - 5 1562 90
XM_005246066.1 XP_005246123.1 2 - 3 683 90
XM_006712253.1 XP_006712316.1 5 - 6 2137 90
XM_011510574.1 XP_011508876.1 5 - 6 2137 90
XM_017003303.1 XP_016858792.1 10 - 11 2187 90
GenBank mRNA AK092021.1 - 3 - 4 90
AK304870.1 - 4 - 5 1497 90
BX647500.1 - 4 - 5 1524 90
X52022.1 - 5 - 6 2150 90

View Full Product Details