Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LETM2
Entrez Gene ID
137994
Gene Name
leucine zipper and EF-hand containing transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.8: 38382505 - 38409527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199659.2 NP_001186588.1 6 - 7 1067 72
NM_001199660.2 NP_001186589.1 6 - 7 906 72
NM_001286787.1 NP_001273716.1 6 - 7 1053 72
NM_001286819.1 NP_001273748.1 6 - 7 1138 72
NM_144652.3 NP_653253.1 5 - 6 923 72
XM_006716291.3 XP_006716354.1 6 - 7 1028 72
XM_011544406.1 XP_011542708.1 6 - 7 1027 72
XM_011544407.1 XP_011542709.1 6 - 7 989 72
XM_011544408.2 XP_011542710.1 6 - 7 3634 72
XM_011544409.2 XP_011542711.1 6 - 7 1307 72
XM_011544410.2 XP_011542712.1 3 - 4 555 72
XM_017013051.1 XP_016868540.1 6 - 7 1370 72
XM_017013052.1 XP_016868541.1 6 - 7 1098 72
XM_017013053.1 XP_016868542.1 6 - 7 3444 72
XM_017013054.1 XP_016868543.1 6 - 7 1083 72
XM_017013055.1 XP_016868544.1 6 - 7 1226 72
XM_017013056.1 XP_016868545.1 6 - 7 1072 72
XM_017013057.1 XP_016868546.1 5 - 6 3488 72
GenBank mRNA AK058138.1 - 5 - 6 862 72
AK302317.1 - 6 - 7 1006 72
AK302474.1 - 6 - 7 878 72
AY173945.1 - 6 - 7 998 72
BC029541.1 - 3 - 4 491 72
BC069830.1 - 6 - 7 987 72

View Full Product Details