Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LETM2
Entrez Gene ID
137994
Gene Name
leucine zipper and EF-hand containing transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.8: 38382505 - 38409527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199659.2 NP_001186588.1 9 - 10 1399 64
NM_001199660.2 NP_001186589.1 9 - 10 1238 64
NM_001286787.1 NP_001273716.1 9 - 10 1385 64
NM_001286819.1 NP_001273748.1 9 - 10 1470 64
NM_144652.3 NP_653253.1 8 - 9 1255 64
XM_006716291.3 XP_006716354.1 9 - 10 1360 64
XM_011544406.1 XP_011542708.1 9 - 10 1359 64
XM_011544407.1 XP_011542709.1 9 - 10 1321 64
XM_011544409.2 XP_011542711.1 9 - 10 1639 64
XM_011544410.2 XP_011542712.1 6 - 7 887 64
XM_017013051.1 XP_016868540.1 9 - 10 1702 64
XM_017013052.1 XP_016868541.1 9 - 10 1430 64
XM_017013053.1 XP_016868542.1 9 - 10 3776 64
XM_017013054.1 XP_016868543.1 9 - 10 1415 64
XM_017013055.1 XP_016868544.1 9 - 10 1558 64
XM_017013056.1 XP_016868545.1 9 - 10 1404 64
XM_017013057.1 XP_016868546.1 8 - 9 3820 64
GenBank mRNA AK058138.1 - 8 - 9 1194 64
AK302317.1 - 9 - 10 1338 64
BC029541.1 - 6 - 7 823 64
BC069830.1 - 9 - 10 1319 64
CB242043.1 - 2 - 3 64

View Full Product Details