The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023010.3 | NP_075386.1 | 4 - 5 | 571 | 111 |
NM_080632.2 | NP_542199.1 | 4 - 5 | 571 | 111 | |
XM_017029737.1 | XP_016885226.1 | 4 - 5 | 571 | 111 | |
XM_017029738.1 | XP_016885227.1 | 4 - 5 | 571 | 111 | |
XM_017029739.1 | XP_016885228.1 | 4 - 5 | 571 | 111 | |
XM_017029740.1 | XP_016885229.1 | 4 - 5 | 571 | 111 | |
GenBank mRNA | AF318576.1 | - | 4 - 5 | 491 | 111 |
AK093728.1 | - | 4 - 5 | 509 | 111 | |
AY013251.1 | - | 4 - 5 | 541 | 111 | |
BC121017.1 | - | 4 - 5 | 514 | 111 | |
BC121018.1 | - | 4 - 5 | 514 | 111 | |
BI549935.1 | - | 4 - 5 | 550 | 110 |