Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMTN
Entrez Gene ID
2039
Gene Name
dematin actin binding protein
Gene Aliases
DMT, EPB49
Chromosome Location
Chr.8: 22048955 - 22082527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114135.4 NP_001107607.1 15 - 16 1375 96
NM_001114136.2 NP_001107608.1 15 - 16 1331 96
NM_001114137.3 NP_001107609.1 14 - 15 1439 96
NM_001114138.2 NP_001107610.1 14 - 15 1426 96
NM_001114139.3 NP_001107611.1 13 - 14 1323 96
NM_001302816.2 NP_001289745.1 14 - 15 1559 96
NM_001302817.2 NP_001289746.1 13 - 14 1344 96
NM_001323378.1 NP_001310307.1 15 - 16 1464 96
NM_001323380.1 NP_001310309.1 15 - 16 1371 96
NM_001323381.1 NP_001310310.1 14 - 15 1480 96
NM_001323382.1 NP_001310311.1 15 - 16 1410 96
NM_001323383.1 NP_001310312.1 14 - 15 1265 96
NM_001323384.1 NP_001310313.1 14 - 15 1398 96
NM_001323385.1 NP_001310314.1 13 - 14 1414 96
NM_001323387.1 NP_001310316.1 13 - 14 1363 96
NM_001323388.1 NP_001310317.1 13 - 14 1211 96
NM_001323389.1 NP_001310318.1 14 - 15 1419 96
NM_001323390.1 NP_001310319.1 14 - 15 1286 96
NM_001323391.1 NP_001310320.1 14 - 15 1389 96
NM_001323392.1 NP_001310321.1 14 - 15 1550 96
NM_001323393.1 NP_001310322.1 14 - 15 1256 96
NM_001323394.1 NP_001310323.1 14 - 15 1417 96
NM_001323395.1 NP_001310324.1 14 - 15 1296 96
NM_001323396.1 NP_001310325.1 14 - 15 1300 96
NM_001323397.1 NP_001310326.1 14 - 15 1468 96
NM_001323398.1 NP_001310327.1 12 - 13 1278 96
NM_001323399.1 NP_001310328.1 12 - 13 1145 96
NM_001323400.1 NP_001310329.1 11 - 12 1294 96
NM_001323401.1 NP_001310330.1 12 - 13 1318 96
NM_001978.4 NP_001969.2 15 - 16 1625 96
XM_005273432.1 XP_005273489.1 15 - 16 1357 96
XM_005273433.1 XP_005273490.1 15 - 16 1289 96
XM_005273442.1 XP_005273499.1 14 - 15 1141 96
XM_011544435.1 XP_011542737.1 14 - 15 1550 96
XM_017013189.1 XP_016868678.1 14 - 15 1232 96
XM_017013190.1 XP_016868679.1 13 - 14 1484 96
XM_017013191.1 XP_016868680.1 13 - 14 1238 96
XM_017013192.1 XP_016868681.1 13 - 14 1119 96
XM_017013193.1 XP_016868682.1 13 - 14 1317 96
XM_017013194.1 XP_016868683.1 13 - 14 1184 96
XM_017013195.1 XP_016868684.1 13 - 14 1484 96
XM_017013196.1 XP_016868685.1 13 - 14 1087 96
XM_017013197.1 XP_016868686.1 13 - 14 1318 96
XM_017013198.1 XP_016868687.1 13 - 14 1160 96
GenBank mRNA AB209718.1 - 11 - 12 851 96
AK055842.1 - 14 - 15 1223 96
AK091581.1 - 13 - 14 1238 96
AK289650.1 - 15 - 16 1257 96
AK295452.1 - 13 - 14 1230 96
BC006318.2 - 15 - 16 1374 96
BC017445.1 - 15 - 16 1374 96
BC052805.1 - 14 - 15 1349 96
BT007396.1 - 14 - 15 1102 96
U28389.1 - 15 - 16 1552 96

View Full Product Details