The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006820.3 | NP_006811.2 | 5 - 6 | 1072 | 119 |
XM_005270391.3 | XP_005270448.1 | 4 - 5 | 579 | 119 | |
XM_005270392.4 | XP_005270449.1 | 3 - 4 | 359 | 119 | |
XM_005270393.4 | XP_005270450.1 | 2 - 3 | 281 | 119 | |
XM_006710304.3 | XP_006710367.1 | 4 - 5 | 386 | 119 | |
XM_011540539.2 | XP_011538841.1 | 5 - 6 | 910 | 119 | |
XM_017000120.1 | XP_016855609.1 | 5 - 6 | 580 | 119 | |
GenBank mRNA | AB000115.1 | - | 5 - 6 | 996 | 119 |
AK223087.1 | - | 5 - 6 | 1006 | 119 | |
AK225442.1 | - | 5 - 6 | 973 | 119 | |
AK293385.1 | - | 5 - 6 | 671 | 119 | |
AK316232.1 | - | 4 - 5 | 521 | 119 | |
AL832618.2 | - | 3 - 4 | 3675 | 119 | |
BC015932.2 | - | 5 - 6 | 978 | 119 |