Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TECPR2
Entrez Gene ID
9895
Gene Name
tectonin beta-propeller repeat containing 2
Gene Aliases
KIAA0329, SPG49
Chromosome Location
Chr.14: 102362912 - 102502481 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172631.2 NP_001166102.1 13 - 14 3353 81
NM_014844.4 NP_055659.2 13 - 14 3353 81
GenBank mRNA AB002295.1 - 10 - 11 2656 81
AB002327.1 - 13 - 14 3302 81
AK299690.1 - 6 - 7 753 81
BC136647.1 - 12 - 13 3150 81
BC142667.1 - 13 - 14 3186 81
BC142715.1 - 13 - 14 3186 81
BC151230.1 - 13 - 14 3326 81

View Full Product Details