Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TECPR2
Entrez Gene ID
9895
Gene Name
tectonin beta-propeller repeat containing 2
Gene Aliases
KIAA0329, SPG49
Chromosome Location
Chr.14: 102362912 - 102502481 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172631.2 NP_001166102.1 5 - 6 916 82
NM_014844.4 NP_055659.2 5 - 6 916 82
GenBank mRNA AB002295.1 - 2 - 3 219 82
AB002327.1 - 5 - 6 865 82
AK127721.1 - 5 - 6 1349 82
BC136647.1 - 4 - 5 713 82
BC142667.1 - 5 - 6 749 82
BC142715.1 - 5 - 6 749 82
BC151230.1 - 5 - 6 889 82

View Full Product Details