The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000288.3 | NP_000279.1 | 4 - 5 | 518 | 72 |
XM_005267019.4 | XP_005267076.1 | 4 - 5 | 798 | 72 | |
XM_017010934.1 | XP_016866423.1 | 4 - 5 | 862 | 72 | |
GenBank mRNA | BC006268.1 | - | 4 - 5 | 460 | 72 |
BC031606.1 | - | 4 - 5 | 502 | 72 | |
CR542054.1 | - | 4 - 5 | 416 | 72 | |
CR542076.1 | - | 4 - 5 | 416 | 72 | |
U76560.1 | - | 4 - 5 | 464 | 72 | |
U88871.1 | - | 4 - 5 | 497 | 72 |