Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX7
Entrez Gene ID
5191
Gene Name
peroxisomal biogenesis factor 7
Gene Aliases
PBD9B, PTS2R, RCDP1, RD
Chromosome Location
Chr.6: 136822220 - 136913934 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000288.3 NP_000279.1 8 - 9 907 72
XM_005267019.4 XP_005267076.1 8 - 9 1187 72
XM_006715502.1 XP_006715565.1 5 - 6 957 72
GenBank mRNA BC006268.1 - 8 - 9 849 72
BC031606.1 - 9 - 10 999 72
CR542054.1 - 8 - 9 805 72
CR542076.1 - 8 - 9 805 72
U76560.1 - 8 - 9 853 72
U88871.1 - 8 - 9 886 72

View Full Product Details