The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031952.3 | NP_114158.2 | 1 - 2 | 218 | 135 |
NR_125327.1 | 1 - 2 | 218 | 135 | ||
NR_125329.1 | 1 - 2 | 218 | 135 | ||
XM_017009948.1 | XP_016865437.1 | 1 - 2 | 199 | 135 | |
GenBank mRNA | AK292338.1 | - | 1 - 2 | 199 | 135 |
AY027526.1 | - | 1 - 2 | 167 | 135 | |
BC032832.1 | - | 1 - 2 | 223 | 135 | |
BC047333.1 | - | 1 - 2 | 204 | 135 |