The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181744.3 | NP_859528.1 | 1 - 2 | 157 | 104 |
XM_017010416.1 | XP_016865905.1 | 1 - 2 | 159 | 104 | |
GenBank mRNA | AY288419.1 | - | 1 - 2 | 129 | 104 |
AY377391.1 | - | 1 - 2 | 134 | 104 | |
BC126194.1 | - | 1 - 2 | 157 | 104 | |
BC126198.1 | - | 1 - 2 | 157 | 104 |