The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016591.2 | NP_057675.1 | 1 - 1 | 3383 | 163 |
GenBank mRNA | AF132035.1 | - | 1 - 1 | 3382 | 163 |