Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NR2C1
Entrez Gene ID
7181
Gene Name
nuclear receptor subfamily 2 group C member 1
Gene Aliases
TR2
Chromosome Location
Chr.12: 95020229 - 95073628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032287.2 NP_001027458.1 6 - 7 949 122
NM_001127362.1 NP_001120834.1 6 - 7 949 122
NM_003297.3 NP_003288.2 6 - 7 949 122
XM_005269133.2 XP_005269190.1 6 - 7 936 122
XM_006719584.2 XP_006719647.1 6 - 7 936 122
XM_006719585.2 XP_006719648.1 7 - 8 974 122
XM_011538716.2 XP_011537018.1 6 - 7 936 122
XM_011538717.2 XP_011537019.1 6 - 7 936 122
GenBank mRNA AB307707.1 - 5 - 6 693 122
AK291319.1 - 6 - 7 913 122
BC026074.1 - 5 - 6 1326 122
BC040141.1 - 6 - 7 901 122
JQ308177.1 - 5 - 6 693 122
JQ308178.1 - 5 - 6 693 122
M21985.1 - 6 - 7 819 122
M29959.1 - 6 - 7 819 122
M29960.1 - 6 - 7 749 122

View Full Product Details