Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF2
Entrez Gene ID
22807
Gene Name
IKAROS family zinc finger 2
Gene Aliases
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location
Chr.2: 212999684 - 213152753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079526.1 NP_001072994.1 3 - 4 354 108
NM_016260.2 NP_057344.2 2 - 3 207 108
XM_005246384.4 XP_005246441.1 2 - 3 317 108
XM_005246385.3 XP_005246442.1 2 - 3 347 108
XM_005246386.4 XP_005246443.1 2 - 3 276 108
XM_011510814.2 XP_011509116.1 2 - 3 681 108
XM_011510819.2 XP_011509121.1 2 - 3 276 108
XM_017003589.1 XP_016859078.1 2 - 3 347 108
XM_017003590.1 XP_016859079.1 2 - 3 317 108
GenBank mRNA AK301503.1 - 2 - 3 203 108
BC028936.1 - 3 - 4 354 108

View Full Product Details