Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HS1BP3
Entrez Gene ID
64342
Gene Name
HCLS1 binding protein 3
Gene Aliases
ETM2, HS1-BP3
Chromosome Location
Chr.2: 20553861 - 20651661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022460.3 NP_071905.3 2 - 3 233 68
XM_017004696.1 XP_016860185.1 2 - 3 236 68
XM_017004697.1 XP_016860186.1 2 - 3 236 68
XM_017004698.1 XP_016860187.1 2 - 3 236 68
XM_017004699.1 XP_016860188.1 2 - 3 288 68
XM_017004700.1 XP_016860189.1 2 - 3 332 68
XM_017004701.1 XP_016860190.1 2 - 3 236 68
GenBank mRNA AK024311.1 - 2 - 3 226 68
AK314383.1 - 2 - 3 229 68
BC050636.1 - 2 - 3 213 68
BX354596.2 - 2 - 3 264 68

View Full Product Details